missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD59 Monoclonal Antibody (MEM-129), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119023
Additional Details : Weight : 0.01000kg
Description
Functional Application: Activation.
CD59 Monoclonal antibody specifically detects CD59 in Human, Porcine samples. It is validated for Flow Cytometry, Functional Assay.Specifications
CD59 | |
Monoclonal | |
1 mg/mL | |
HEPES buffered saline with no preservative; pH 7.0 | |
O62680, P13987 | |
CD59 | |
Human peripheral blood lymphocytes. | |
100 μg | |
Primary | |
Human, Porcine | |
Antibody | |
IgM |
Flow Cytometry, Functional Assay | |
MEM-129 | |
Unconjugated | |
CD59 | |
16.3A5; 1F5; 1F5 antigen; 20 kDa homologous restriction factor; AA987121; Cb59b molecule; CD59; CD59 antigen; CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, EJ30, EL32 and G344); CD59 glycoprotein; CD59 molecule; CD59 molecule (CD59 blood group); CD59 molecule, complement regulatory protein; Cd59a; CD59a antigen; CD59A glycoprotein; Cd59b; CD59b antigen; CD59B glycoprotein; CD59b moleucle, complement regulatory protein; complement inhibitor CD59; complement regulatory protein; EJ16; EJ30; EL32; G344; HRF20; HRF-20; human leukocyte antigen MIC11; Ly-6-like protein; lymphocytic antigen CD59/MEM43; MACIF; MAC-inhibitory protein; MACIP; MAC-IP; MEM43; MEM43 antigen; membrane attack complex (MAC) inhibition factor; membrane attack complex inhibition factor; Membrane inhibitor of reactive lysis; MIC11; MIN1; MIN2; MIN3; MIRL; MSK21; p18-20; protectin; surface anitgen recognized by monoclonal antibody 16.3A5; T cell-activating protein | |
Mouse | |
sequential chromatography | |
RUO | |
397347, 966 | |
4° C, do not freeze | |
Liquid |