missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD56 Monoclonal Antibody (NKI-nbl-1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA135251
Additional Details : Weight : 0.01000kg
Description
In IHC applications, this antibody only works on frozen sections and not on paraffin sections.
CD56 Monoclonal antibody specifically detects CD56 in Human samples. It is validated for Immunohistochemistry (Frozen).Specifications
CD56 | |
Monoclonal | |
Conc. Not Determined | |
ascites diluted in PBS with 0.02% sodium azide | |
P13591 | |
Ncam1 | |
NCAM. | |
RUO | |
4684 | |
4° C | |
Liquid |
Immunohistochemistry (Frozen) | |
NKI-nbl-1 | |
Unconjugated | |
Ncam1 | |
adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; Cd56; CD-56; CD56 120 kDa GPI-linked isoform; CD56 140 kDa isoform; CD56 140 kDa VASE isoform; E NCAM; E-NCAM; MSK39; N CAM1; NCAM; N-CAM; Ncam1; N-CAM-1; NCAM-1; NCAMC; NCAM-C; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM | |
Mouse | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG1 |