missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD55 Monoclonal Antibody (MEM-118), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119573
Additional Details : Weight : 0.01000kg
Description
CD55 Monoclonal antibody specifically detects CD55 in Human, Non-human Primate samples. It is validated for Flow Cytometry.Specifications
CD55 | |
Monoclonal | |
FITC | |
CD55 | |
CD55; CD55 antigen; CD55 molecule (Cromer blood group); CD55 molecule, decay accelerating factor for complement; CD55 molecule, decay accelerating factor for complement (Cromer blood group); Cd55a; Complement decay-accelerating factor; complement decay-accelerating factor, GPI-anchored; complement-glycosylphosphatidylinositol; CR; CROM; Cromer blood group; Daf; Daf1; DAF-GPI; decay accelarating factor 1; decay accelerating factor 1; decay accelerating factor GPI-form; decay-accelarating factor; glycosylphophatidylinositol-anchored form; GPI anchor addition signal; GPI-DAF; RP11-357P18.1; TC | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
1604 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-118 | |
TBS with 0.2% BSA and 15mM sodium azide; pH 8.0 | |
P08174 | |
CD55 | |
T cell line HPB-ALL. | |
100 Tests | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgM |