missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 Monoclonal Antibody (QBEND/10), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110204
246.01 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
This antibody will not cross-react with bovine, canine or sheep.
CD34 Monoclonal antibody specifically detects CD34 in Human, Non-human Primate samples. It is validated for Flow Cytometry.Specifications
CD34 | |
Monoclonal | |
FITC | |
Cd34 | |
(E3-independent) E2 ubiquitin-conjugating enzyme R1; AU040960; CD34; CD34 antigen; CD34 molecule; CDC34; cell division cycle 34; cell division cycle 34 homolog; cluster designation 34; cluster of differentiation antigen 34; E2 ubiquitin-conjugating enzyme R1; E2-CDC34; hematopoietic progenitor cell antigen CD34; hematopoietic progenitor cell marker CD34; Mucosialin; RP11-328D5.2; sialomucin protein; type 1 transmembrane protein; UBC3; UBCH3; UBE2R1; ubiquitin carrier protein; ubiquitin conjugating enzyme E2 R1; ubiquitin-conjugating enzyme E2 R1; Ubiquitin-conjugating enzyme E2-32 kDa complementing; ubiquitin-conjugating enzyme E2-CDC34; Ubiquitin-protein ligase R1 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
947 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
QBEND/10 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P28906 | |
Cd34 | |
Human endothelial vesicles | |
100 Tests | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |