Learn More
CD33 Monoclonal Antibody (WM53), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516982
Additional Details : Weight : 0.01000kg
Description
Overnight incubation is recommended. The epitope recognized by this antibody is reported to be sensitive to formaldehyde fixation and tissue processing. We recommend the use of acetone fixation for frozen sections. A suggested positive control for immunohistochemical applications is lymphatic tissue. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Overnight incubation with primary antibody is recommended.
CD33 Monoclonal antibody specifically detects CD33 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blot.Specifications
CD33 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P20138 | |
CD33 | |
Human AML cells | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunoprecipitation, Western Blot | |
WM53 | |
Unconjugated | |
CD33 | |
CD33; CD33 antigen; CD33 antigen (gp67); CD33 molecule; CD33 molecule transcript; FLJ00391; gp67; Myeloid cell surface antigen CD33; p67; sialic acid binding Ig-like lectin 3; sialic acid-binding Ig-like lectin 3; SIGLEC3; Siglec-3 | |
Mouse | |
Protein G | |
RUO | |
945 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |