missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FCGR2B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527138
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: THP-1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FCGR2B Polyclonal antibody specifically detects FCGR2B in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
FCGR2B | |
Polyclonal | |
Unconjugated | |
FCGR2B | |
AI528646; CD32; CD32 receptor 2; CD32 receptor 2a; CD32 receptor 2b; CD32A; CD32B; CDw32; F630109E10Rik; Fc fragment of IgG receptor IIb; Fc fragment of IgG, low affinity II, receptor for (CD32); Fc fragment of IgG, low affinity IIb, receptor; Fc fragment of IgG, low affinity IIb, receptor (CD32); Fc fragment of IgG, low affinity IIb, receptor for (CD32); fc gamma receptor IIB; Fc gamma RIIb; Fc receptor, IgG, low affinity IIb; Fc[g]RII; FCG2; FC-gamma RII; fc-gamma RII-a; Fc-gamma RII-b; Fc-gamma-RII; fc-gamma-RIIa; FcgammaRIIB; fc-gamma-RIIb; FcgammaRIIB2; FcGR; Fcgr2; Fcgr2a; FCGR2A1; FCGR2B; FcgRII; Fcr-2; Fcr-3; fcRII; fcRII-a; fcRII-b; FGFR2B; IGFR2; IgG Fc receptor II beta; igG Fc receptor II-b; Low affinity immunoglobulin gamma Fc region receptor II; low affinity immunoglobulin gamma Fc region receptor II-b; Ly-17; LyM-1; Ly-m20; Lymphocyte antigen 17 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2213 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
P31994 | |
FCGR2B | |
Synthetic peptide corresponding to a region within amino acids 248 and 310 of Human CD32B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |