Learn More
FCGR2B Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547122
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) Fc gamma RIIA is observed, and approximately 10% cross-reactivity with recombinant mouse Fc gamma RIIB is observed, and less than 1% cross-reactivity with rhFc gamma RIIIA is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
FCGR2B Polyclonal antibody specifically detects FCGR2B in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
FCGR2B | |
Polyclonal | |
Unconjugated | |
FCGR2B | |
AI528646; CD32; CD32 receptor 2; CD32 receptor 2a; CD32 receptor 2b; CD32A; CD32B; CDw32; F630109E10Rik; Fc fragment of IgG receptor IIb; Fc fragment of IgG, low affinity II, receptor for (CD32); Fc fragment of IgG, low affinity IIb, receptor; Fc fragment of IgG, low affinity IIb, receptor (CD32); Fc fragment of IgG, low affinity IIb, receptor for (CD32); fc gamma receptor IIB; Fc gamma RIIb; Fc receptor, IgG, low affinity IIb; Fc[g]RII; FCG2; FC-gamma RII; fc-gamma RII-a; Fc-gamma RII-b; Fc-gamma-RII; fc-gamma-RIIa; FcgammaRIIB; fc-gamma-RIIb; FcgammaRIIB2; FcGR; Fcgr2; Fcgr2a; FCGR2A1; FCGR2B; FcgRII; Fcr-2; Fcr-3; fcRII; fcRII-a; fcRII-b; FGFR2B; IGFR2; IgG Fc receptor II beta; igG Fc receptor II-b; Low affinity immunoglobulin gamma Fc region receptor II; low affinity immunoglobulin gamma Fc region receptor II-b; Ly-17; LyM-1; Ly-m20; Lymphocyte antigen 17 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
2213 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P31994 | |
FCGR2B | |
Mouse myeloma cell line NS0-derived recombinant human Fc gamma RII/CD32 Ala46-Pro217 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |