missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD31 Monoclonal Antibody (MEM-05), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119769
Additional Details : Weight : 0.01000kg
Description
CD31 Monoclonal antibody specifically detects CD31 in Human samples. It is validated for Flow Cytometry.Specifications
CD31 | |
Monoclonal | |
PE | |
Pecam1 | |
C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
5175 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-05 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P16284 | |
Pecam1 | |
Leukocytes of patient suffering from LGL-type leukaemia. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |