Learn More
CD31 Monoclonal Antibody (2F7B2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515336
Additional Details : Weight : 0.01000kg
Description
MA5-15336 targets CD31 in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15336 immunogen is purified recombinant fragment of human CD31 expressed in E. Coli.
CD31 Monoclonal antibody specifically detects CD31 in Human samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
CD31 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P16284 | |
Pecam1 | |
Purified recombinant fragment of human CD31 expressed in E. Coli. | |
RUO | |
5175 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
2F7B2 | |
Unconjugated | |
Pecam1 | |
C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |