Learn More
CD31 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA137326
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse samples based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is tonsil tissue.
CD31 Polyclonal antibody specifically detects CD31 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
CD31 | |
Polyclonal | |
Unconjugated | |
Pecam1 | |
C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5175 | |
4° C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.013 mg/mL | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.2 | |
P16284 | |
Pecam1 | |
Synthetic peptide corresponding to C-terminus of mouse CD31 protein. | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG |