Learn More
CD200R Monoclonal Antibody (OX-110), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA182801
Additional Details : Weight : 0.01000kg
Description
This product is a low-endotoxin, preservative-free formulation. Endotoxin level is <0.01EU/ug. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Rat anti Mouse CD200R antibody, clone OX-110 recognizes mouse CD200R, a cell surface glycoprotein (also known as OX2R.
CD200R Monoclonal antibody specifically detects CD200R in Mouse samples. It is validated for Flow Cytometry.Specifications
CD200R | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9ES57 | |
CD200R1 | |
Fusion protein mCD200RCD4d3+4. | |
500 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry | |
OX-110 | |
Unconjugated | |
CD200R1 | |
antigen identified by monoclonal antibody MRC OX-2 receptor; CD200 cell surface glycoprotein receptor; CD200 R1; CD200 receptor 1; CD200R; Cd200r1; Cell surface glycoprotein CD200 receptor 1; cell surface glycoprotein OX2 receptor 1; cell surface glycoprotein receptor CD200; CRTR2; HCRTR2; MOX2 receptor; MOX2R; Orexin receptor 2; OX102 antigen; OX2 receptor; OX2R; UNQ2522/PRO6015 | |
Rat | |
Protein G | |
RUO | |
57781 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |