missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD178 Monoclonal Antibody (GM5F4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA117757
Additional Details : Weight : 0.01000kg
Description
This antibody recognizes native human FasL protein expressed in transfected mammalian cells.
CD178 Monoclonal antibody specifically detects CD178 in Human samples. It is validated for ELISA, Flow Cytometry.Specifications
CD178 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.01% sodium azide; pH 7.2 | |
P48023 | |
Faslg | |
cDNA encoding human FasL (extracellular domain) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry | |
GM5F4 | |
Unconjugated | |
Faslg | |
ADAM10-processed FasL form; ALPS1B; APL; Apoptosis (APO 1) antigen ligand 1; apoptosis (APO-1) antigen ligand 1; Apoptosis (APO-1) antigen ligand 1 (Fas antigen ligand); apoptosis antigen ligand; APT1LG1; APTL; CD178; CD178 antigen; CD95 ligand; CD95 ligand protein;Generalized lymphoproliferative disease (Gld); Cd95l; CD95-L; CD95L protein; fas antigen ligand; Fas ligand; Fas ligand (TNF superfamily, member 6); Fasl; Fas-L; FasL ICD; FasL intracellular domain; Faslg; Fas-LG; generalized lymphoproliferative disease; gld; mutant tumor necrosis factor family member 6; Receptor-binding FasL ectodomain; sFasL; Soluble Fas ligand; soluble form; SPA; SPPL2A-processed FasL form; TNFL; Tnfsf6; TNLG1A; Tumor necrosis factor (ligand) superfamily member 6; Tumor necrosis factor (ligand) superfamily member 6 (apoptosis (APO-1) antigen ligand 1) (Fas antigen ligand); Tumor necrosis factor (ligand) superfamily, member 6 (apoptosis (APO-1) antigen ligand 1) (Fas antigen ligand); Tumor necrosis factor | |
Mouse | |
Protein G | |
RUO | |
356 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |