missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRP alpha Monoclonal Antibody (MRC OX-41), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA190103
Additional Details : Weight : 0.01000kg
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SIRP alpha Monoclonal antibody specifically detects SIRP alpha in Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Radioimmune Assays (RIA).Specifications
SIRP alpha | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.5% BSA and 0.02% sodium azide | |
P97710 | |
SIRPA | |
Rat peritoneal macrophages | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Radioimmune Assays (RIA) | |
MRC OX-41 | |
FITC | |
SIRPA | |
AI835480; Bit; brain Ig-like molecule with tyrosine-based activation motifs; Brain immunoglobulin like protein with tyrosine - based activation motifs; brain immunological-like with tyrosine-based motifs; brain-immunoglobulin-like molecule with tyrosine-based activation motifs; CD172; CD172 alpha; CD172 antigen-like family member A; CD172A; Inhibitory receptor SHPS-1; macrophage fusion receptor; Macrophage membrane protein MFP150; MFR; mSIRP-alpha1; Myd1; MYD-1; myD-1 antigen; p84; Protein tyrosine phosphatase non-receptor type substrate 1 (SHP substrate 1); protein tyrosine phosphatase, non-receptor type substrate 1; Protein tyrosine phosphatase, non-receptor type substrate 1 (SHP substrate 1); Ptpns1; SHP substrate 1; SHP-1; SHPS1; SHPS-1; signal regulatory protein alpha; signal-regulatory protein alpha; signal-regulatory protein alpha-1; Signal-regulatory protein alpha-2; Signal-regulatory protein alpha-3; SIRP; Sirpa; SIRPalpha; Sirp-alpha-1; SIRPalpha2; Sirp-alpha-2; Sirp-alpha-3; | |
Mouse | |
IgG fraction | |
RUO | |
25528 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |