Learn More
CD169 Monoclonal Antibody (ED3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180164
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti rat CD169 antibody, clone ED3 recognizes the rat CD169 cell surface antigen, a approximately 185 kDa molecule expressed by macrophages, predominately confined to lymphoid organs only.
CD169 Monoclonal antibody specifically detects CD169 in Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation.Specifications
CD169 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
0 | |
SIGLEC1 | |
Rat Spleen cell homogenate. | |
250 μg | |
Primary | |
Rat | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation | |
ED3 | |
Unconjugated | |
SIGLEC1 | |
Cd169; dJ1009E24.1; macrophage receptor; p210; pSn; SA; SER; sheep erythrocyte receptor; sialic acid binding Ig like lectin 1; sialic acid binding Ig-like lectin 1, sialoadhesin; Sialic acid-binding Ig-like lectin 1; sialic acid-binding immunoglobulin-like lectin 1; sialoadhesin; SIGLEC1; Siglec-1; SN | |
Mouse | |
Protein A | |
RUO | |
311426 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |