missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163 Monoclonal Antibody (215930), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523788
Additional Details : Weight : 0.00010kg
Description
In sandwich ELISAs, no cross-reactivity or interference with recombinant mouse (rm) CD163, rmCD6, or recombinant human CD6 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
CD163 Monoclonal antibody specifically detects CD163 in Human samples. It is validated for ELISA.Specifications
CD163 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q86VB7 | |
CD163 | |
Mouse myeloma cell line NS0-derived recombinant human CD163 Gly41-Ser1045 | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
215930 | |
Unconjugated | |
CD163 | |
CD163; CD163 antigen; CD163 molecule; CD163v2; CD163v3; Hemoglobin scavenger receptor; M130; macrophage-associated antigen; MM130; putative CD163 antigen; SCARI1; Scavenger receptor cysteine-rich type 1 protein M130; sCD163; Soluble CD163; Soluble sCD163 | |
Mouse | |
Protein A/G | |
RUO | |
9332 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |