missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR2DL4 Monoclonal Antibody (mAb#33), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110143
Additional Details : Weight : 0.01000kg
Description
This antibody recognizes the extracellular portion of CD158d.
KIR2DL4 Monoclonal antibody specifically detects KIR2DL4 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
KIR2DL4 | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Q99706 | |
KIR2DL4 | |
NK3.3 cells and KIR2DL4-Ig fusion protein | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
mAb#33 | |
Unconjugated | |
KIR2DL4 | |
CD158 antigen-like family member D; CD158D; G9P; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 4; killer cell immunoglobulin-like receptor 2DL4; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4; killer cell inhibitory receptor 103AS; killer Ig receptor; killer-cell immunoglobulin-like receptor; killer-cell immunoglobulin-like receptor 2DL4; KIR; KIR103; KIR103AS; KIR-103AS; KIR2DL4; KIR-2DL4; MHC class I NK cell receptor KIR103AS; natural killer cell inhibitory receptor; natural killer cell receptor; NK cell receptor | |
Mouse | |
Protein A | |
RUO | |
3805 | |
4° C, do not freeze | |
Liquid |