missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BST-1 Monoclonal Antibody (KT157), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA517947
Additional Details : Weight : 0.01000kg
Description
MA5-17947 targets CD157/BST1 in FACS applications and shows reactivity with Mouse samples. The MA5-17947 immunogen is t cell hybridoma J774.
Bone marrow stromal cell antigen 1 (BST1) is a stromal cell line-derived glycosylphosphatidylinositol-anchored molecule that facilitates pre-B cell growth. BST1, which is also known as ADP-rybosyl cyclase 2 and DAMP-1, is an enzyme which is expressed by BST1 gene in human. BST1 expression is enhanced in bone marrow stromal cell lines derived from patients with rheumatoid arthritis. The polyclonal B cell abnormalities in rheumatoid arthritis may be, at least in part, attributed to BST1 overexpression in the stromal cell population.Specifications
BST-1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q64277 | |
Bst1 | |
T cell hybridoma J774 | |
250 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2c |
Flow Cytometry | |
KT157 | |
Unconjugated | |
Bst1 | |
114/A10; A530073F09; ADP-ribosyl cyclase 2; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 2; antigen BP3; Bone marrow stromal antigen 1; bone marrow stromal cell antigen 1; Bp3; Bp-3; BP-3 alloantigen; Bst1; BST-1; Bsta1; cADPr hydrolase 2; CD157; CD157 antigen; Cyclic ADP-ribose hydrolase 2; leukocyte antigen 65; Ly65; Ly-65; NAD(+) nucleosidase; NAD+ nucleosidase | |
Rat | |
Protein G | |
RUO | |
12182 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |