missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD154 (CD40 Ligand) Monoclonal Antibody (39H5), FITC, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Invitrogen MA516506
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD154 (CD40 Ligand) Monoclonal antibody specifically detects CD154 (CD40 Ligand) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD154 (CD40 Ligand) | |
Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
P27548 | |
CD40LG | |
L cells transfected with CD40L | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Flow Cytometry | |
39H5 | |
FITC | |
CD40LG | |
CD154; CD40 antigen ligand; CD40 ligand; CD40 ligand CD154; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40L; CD40-L; Cd40lg; gp39; hCD40L; H-CD-40-L; HIGM1; IGM; IMD3; Ly62; Ly-62; M-CD-40-L; RP23-153G22.3; T-B cell-activating molecule; T-BAM; T-cell antigen Gp39; TNF-related activation protein; TNFSF5; TRAP; tumor necrosis factor (ligand) superfamily member 5; tumor necrosis factor (ligand) superfamily, member 5; tumor necrosis factor ligand superfamily member 5 | |
Armenian Hamster | |
Protein G | |
RUO | |
21947 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |