missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C/EBP zeta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA512304
Additional Details : Weight : 0.01000kg
Description
C/EBP zeta Polyclonal antibody specifically detects C/EBP zeta in Human samples. It is validated for Western Blot.Specifications
C/EBP zeta | |
Polyclonal | |
Unconjugated | |
CEBPZ | |
AI848081; C EBP; C/EBP zeta; CBF; Cbf2; CCAAT/enhancer binding protein (C/EBP), zeta; CCAAT/enhancer binding protein alpha (C/EBP) related sequence 1; CCAAT/enhancer binding protein cebp family zeta; CCAAT/enhancer binding protein zeta; CCAAT/enhancer-binding protein zeta; CCAAT-binding factor; CCAAT-box-binding transcription factor; CEBP zeta; Cebpa-rs1; CEBPZ; core-binding factor; CTF2; HSPCBF; HSP-CBF; NOC1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
10153 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
Q03701 | |
CEBPZ | |
KLH conjugated synthetic peptide between 1001-1029 amino acids from the C-terminal region of human CBF (CEBPZ) | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |