Learn More
BOLL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530808
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: NT2D1. Predicted reactivity: Mouse (95%), Rat (95%), Rhesus Monkey (100%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BOLL Polyclonal antibody specifically detects BOLL in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
BOLL | |
Polyclonal | |
Unconjugated | |
BOLL | |
4930554P13Rik; 4930597B14Rik; bol, boule-like; bol, boule-like (Drosophila); Boll; BOULE; boule homolog, RNA binding protein; boule-like RNA binding protein; boule-like RNA-binding protein; protein boule-like; RGD1559527 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
66037, 75388 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8N9W6, Q924M5 | |
BOLL | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 167 of Human BOLL | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |