Learn More
Labnet™ Anodized Aluminum Block
Click to view available options
Holds:
12 x 17mm Tubes
Specifications
For Use With (Equipment) | Test Tubes |
Holds | 12 x 17mm Tubes |
Material | Anodized Aluminum |
Type | Block |
Weight (Metric) | 1 kg |