missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sodium Phosphate Dibasic Tetrahydrate, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
6 x 500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 500 g |
Linear Formula | NaNH4HPO4 · 4H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 209.07g/mol |
Percent Purity | ≥99% |
Grade | puriss. |
Chemical Name or Material | Ammonium Sodium Phosphate Dibasic Tetrahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'