Learn More
Ammonium Phosphate Dibasic (Colorless Crystals), Fisher BioReagents
Brand: Fisher Bioreagents BP361-500
Additional Details : CAS Number : 7783-28-0 Weight : 0.55000kg
Description
Poly Bottle; 500g
Ammonium Phosphate Dibasic is suitable for use in many biological applications.Specifications
Ammonium Phosphate Dibasic | |
White | |
Crystalline Solid | |
98.0 to 101.0 % | |
(NH4)2HPO4 | |
15, 540 | |
MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
phosphoric acid diamine | |
24540 | |
132.06 | |
Pass Test | |
1.619g/cm³ |
7783-28-0 | |
7.9 to 8.3 | |
500 g | |
H9N2O4P | |
MFCD00010891 | |
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
N.N.OP(O)(O)=O | |
132.06 | |
CHEBI:63051 | |
98.0 to 101.0% | |
Poly Bottle |