missing translation for 'onlineSavingsMsg'
Learn More

Ammonium acetate, 97%, Thermo Scientific Chemicals

Catalog Number p-7023189
Click to view available options
Quantity:
250 g
1000 g
5000 g
631-61-8
C2H7NO2
77.083
MFCD00013066
USFZMSVCRYTOJT-UHFFFAOYSA-N
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
517165
CHEBI:62947
azanium;acetate
CC(=O)[O-].[NH4+]
631-61-8
C2H7NO2
77.083
MFCD00013066
USFZMSVCRYTOJT-UHFFFAOYSA-N
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
517165
CHEBI:62947
azanium;acetate
CC(=O)[O-].[NH4+]

Ammonium acetate is widely utilized as a catalyst in the Knoevenagel condensation. It is the primary source of ammonia in the Borch reaction in organic synthesis. It is used with distilled water to make a protein precipitating reagent. It acts as a buffer for electrospray ionization (ESI) mass spectrometry of proteins and other molecules and as mobile phases for high performance liquid chromatography (HPLC). Sometimes, it is used as a biodegradable de-icing agent and an acidity regulator in food additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium acetate is widely utilized as a catalyst in the Knoevenagel condensation. It is the primary source of ammonia in the Borch reaction in organic synthesis. It is used with distilled water to make a protein precipitating reagent. It acts as a buffer for electrospray ionization (ESI) mass spectrometry of proteins and other molecules and as mobile phases for high performance liquid chromatography (HPLC). Sometimes, it is used as a biodegradable de-icing agent and an acidity regulator in food additives.

Solubility
Soluble in water, methanol, acetone and liquid ammonia.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY
Melting Point ∼114°C (decomposition)
Density 1.17
Boiling Point (decomposition)
Flash Point 136°C (277°F)
Odor Ammonia-like
Linear Formula CH3COONH4
Quantity 250 g
Beilstein 4186741
Sensitivity Hygroscopic
Merck Index 14,495
Solubility Information Soluble in water,methanol,acetone and liquid ammonia.
Formula Weight 77.08
Percent Purity 97%
Chemical Name or Material Ammonium acetate
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium acetate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'