Learn More
Cytiva Amersham™ ECL™ Direct Nucleic Acid Labeling/Detection System
Use for fast and easy detection of medium-to-high-target amounts. Cytiva Life Sciences™ Amersham™ ECL™ Direct Nucleic Acid Labeling/Detection System are based on the direct labeling of DNA or RNA probes with horseradish peroxidase (HRP) in a simple 20 min chemical reaction
Brand: Cytiva RPN2105
687.98 EUR valid until 2024-12-31
Use promo code "21954" to get your promotional price.
Additional Details : Weight : 0.58092kg
Description
- Direct probe labeling in a 10 min reaction, 1 h from hybridization to detection with ECL Direct, Hybond N+, and Hyperfilm ECL.
- Eliminates handling, waste, and regulatory issues associated with the use of radioactivity.
- No need to strip blots before reprobing.
- For fast and easy detection of medium- to high-target amounts in applications such as colony/plaque screens, dot blots, and PCR product analyses.
- Consistent results combining strong signals with very low backgrounds.
Specifications
2°C to 8°C | |
ECL Detection Reagents 1 and 2, for 4000cm2 membrane | |
For 4000 cm2 membrane |
Colony or plaque screens, dot blots and PCR product analyses | |
ECL Detection Reagents | |
20 min. |