Learn More
Invitrogen™ Ambion™ T7 RNA Polymerase (cloned) 20U/μL
Brand: Invitrogen™ AM2082
Weitere Details : Weight : 0.01000kg
Spezifikation
One tube containing 5000 units at a concentration of 20U/μL. Store at -20deg.C | |
miRNA analysis | |
DNA |
5000 units | |
20U/μL |