Learn More
Invitrogen™ Microcentrifuge Spin Columns
The microcentrifuge spin columns included with the FastTrack™ 2.0 and Micro-FastTrack 2.0 Kits are available separately.
Brand:  Invitrogen™ R50020
1453.50 EUR valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Additional Details : Weight : 0.20000kg
Product Code. 12020196
  • 1938.00 EUR / Pack of 200
Estimated Shipment: 06-06-2024
Description
The microcentrifuge spin columns included with the FastTrack™ 2.0 and Micro-FastTrack 2.0 Kits are available separately. The columns are made from polypropylene and are designed to enhance the speed and efficiency of polyA+ RNA isolation. The columns are sterilized and RNase-free.
Specifications
Specifications
0.45 micron cellulose acetate microcentrifuge spin columns and sterile microcentrifuge tubes. After opening, store microcentrifuge columns and tubes in a sterile container.
200 Columns