Learn More
Amberlite™ IRA-400(Cl), ion exchange resin, Thermo Scientific™
Catalog Number p-4408668
Click to view available options
Quantity:
500 g
2 kg
9002-24-8
Styrene-DVB; Strongly basic gel type resin
MFCD00132708
9002-24-8
Styrene-DVB; Strongly basic gel type resin
MFCD00132708
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Quantity 500 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlite IRA-400, ion exchange resin