Learn More
Fisherbrand™ Amber Wide Mouth Straight Sided Glass Jars with White Polypropylene Caps
GREENER_CHOICE
Product Code. p-7144758 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
120 mL
240 mL
Unit Size:
Pack of 12
Pack of 24
Product Code. 16114587
Only null left
Add to basket
Add to basket
Amber USP Type III soda-lime glass wide mouth design is great for large solid samples and allow for complete removal of contents. Ideal for soil sampling and environmental applications
Designed without shoulders for maximum storage capacity. Wide mouth for easy filling
Specifications
Capacity (Metric) 120 mL
Capacity (English) 4 oz.
Color Amber
Diameter (Metric) Outer Neck 54 mm
Diameter (Metric) Inner Neck 40 mm
For Use With (Application) Soil Sampling and Environmental Applications
Includes Cap
Material Glass
Quantity 24
Closure Color White
Neck Finish 58-400
Certifications/Compliance USP Type III Soda Lime Glass Wide Mouth Design is Great for Large Solid Samples and Allow for Complete Removal of Contents
Closure Included Yes
Mouth Wide
Shape Round
Height (Metric) 69 mm
Product Type Straight-sided Jar
Closure Liner PTFE/Polyethylene Foam
Closure Material Polypropylene
Closure Size 58-400 mm
Diameter (Metric) 60 mm
Closure Type Cap