missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Silicon slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.5 in.) length, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 15443665
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
11145-30-5
AlSi
55.067
MFCD00143595
CSDREXVUYHZDNP-UHFFFAOYSA-N
silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic
9793685
aluminum;silicon
[Al].[Si]
This item is not returnable. View return policy
11145-30-5
AlSi
55.067
MFCD00143595
CSDREXVUYHZDNP-UHFFFAOYSA-N
silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic
9793685
aluminum;silicon
[Al].[Si]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Avg. Mol. Wt. or Mol. Wt. Range Al:Si; 96.5:3.5 approximate wt%
Quantity 100 g
Percent Purity 99.999%
Grade Puratronic™
Form Slug
Assay (metals basis)
Chemical Name or Material Aluminum Silicon slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.5 in.) length

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'