Learn More
VACUUBRAND™ Aluminum Reducing Pieces With Small KF Flanges
48.80 EUR - 63.80 EUR
Specifications
For Use With | Solenoid-operated valves VV and VV C |
---|---|
Material | Aluminum |
Quantity | 1 Each |
Size | 40 mm |
Type | Reducing Pieces |
Product Code | Brand | Diameter (Metric) Inside | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Price | Quantity & Availability | |||||
10029440
|
VACUUBRAND™
20669043 |
40/16 mm |
60.00 EUR
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
10099600
|
VACUUBRAND™
20669041 |
25/16 mm |
48.80 EUR
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
10653952
|
VACUUBRAND™
20669044 |
40/25 mm |
63.80 EUR
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
10707841
|
VACUUBRAND™
20669040 |
25/10 mm |
50.95 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
10593942
|
VACUUBRAND™
20669042 |
40/10 mm |
60.00 EUR
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Specifications
Solenoid-operated valves VV and VV C | |
1 Each | |
Reducing Pieces |
Aluminum | |
40 mm |