Learn More
Welch™ Aluminum KF Inner Centering with Buna N O-ring
6.15 EUR - 9.65 EUR
Product Code | Brand | Diameter (Metric) Outside | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outside | Price | Quantity & Availability | |||||
10762152
|
Welch™
701081 |
16 mm |
6.15 EUR
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
10640153
|
Welch™
701083 |
25 mm |
8.37 EUR
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
10596292
|
Welch™
701085 |
40 mm |
9.65 EUR
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Specifications
Aluminum | |
1 Each | |
KF Inner Centering |