Learn More
Fisherbrand™ Gel Casting Trays for TVG-SYS Vari-gel MIDI Horizontal Gel Electrophoresis System
Accessory for use with TVG-SYS Vari-gel MIDI Horizontal Gel Electrophoresis System.
Brand: Fisherbrand™
Additional Details : Weight : 0.80000kg
Description
- UV transparent gel trays allow for placement directly onto a UV transilluminator for gel viewing once stained
- Asymmetric electrode offset - prevents accidental polarity reversal ensuring no possibility of sample loss into the buffer
- Aluminium casting gates - ensure easy casting of the gel inside or outside of the tank.They slide into pre-formed grooves at the end of the gel tray providing a secure seal against gel leakage
- Available in three different dimensions
Specifications
Gel Casting Tray | |
100 mm | |
100 mm |