missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyltrimethylsilane, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Density | 0.7190g/mL |
Boiling Point | 85.0°C |
Flash Point | 7°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Linear Formula | H2C=CHCH2Si(CH3)3 |
Refractive Index | 1.4060 to 1.4080 |
Quantity | 10 mL |
Show More |
Safety and Handling
Product Identifier
- Allyltrimethylsilane
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 2
- H411-Toxic to aquatic life with long lasting effects.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'