Learn More
Allyl vinyl ether, 95% stab. with 0.1% potassium hydroxide
Product Code. 11358036
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358036 50 g 50g
11348036 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11358036 Supplier Thermo Scientific Alfa Aesar Supplier No. L13320.18
Request Bulk or Custom Format
CAS: 3917-15-5 | C5H8O | 84.12 g/mol
Allyl vinyl ether is used as a intermediate in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Allyl vinyl ether is used as a intermediate in chemical research.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, ultraviolet radiation and free radical initiators.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3917-15-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00039827
InChI Key ZXABMDQSAABDMG-UHFFFAOYSA-N
Synonym allyl vinyl ether, ether, allyl vinyl, vinyl allyl ether, 1-propene, 3-ethenyloxy, allyl ethenyl ether, unii-fj2j0n55cs, fj2j0n55cs, allylvinylether, acmc-20alul, 3-ethenoxy-1-propene
PubChem CID 221523
IUPAC Name 3-ethenoxyprop-1-ene
SMILES C=CCOC=C
Specifications
Density 0.781
Boiling Point 65°C to 68°C
Flash Point <−5°C (23°F)
Odor Characteristic
Linear Formula CH2=CHCH2OCH=CH2
Refractive Index 1.412
Quantity 50 g
UN Number UN3271
Beilstein 1697665
Solubility Information Slightly soluble in water.
Formula Weight 84.12
Percent Purity 95%
Chemical Name or Material Allyl vinyl ether, stabilized with 0.1% potassium hydroxide