Learn More
Alfa Aesar™ Alloferon 1
For cellular and molecular biology applications
Brand: Alfa Aesar™ J66879.M¬
Additional Details : CAS Number : 347884-61-1 Weight : 0.00001kg
Specifications
HPLC | |
Solid | |
Alloferon 1 | |
RUO - research use only |
-30°C to -10°C | |
1265.31 | |
1 mg | |
His-Gly-Val-Ser-Gly-His-Gly-Gln-His-Gly-Val-His-Gly |
For Research Use Only