Learn More
Thin Insulating Tubes, Thermo Scientific™
Oxide ceramics tubes resistant to corrosive liquids and gases
24.80 EUR - 54.35 EUR
Specifications
Diameter (Metric) Inside 1.5mm
Diameter (Metric) Outside 2mm
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11373887
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031489.DM
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11353887
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031489.DR
200 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11347518
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031489.DW
300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Inert to oxidation
  • Not subject to radiation damage
  • Wear resistant and hard
  • With both ends open
Specifications
Specifications
1.5mm
2mm