Learn More
Al-23 Crucible, Cylindrical, Flat Base;Height x OD x ID (mm), 15 x 10 x 8;Vol (ml), 0.7, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 032950.KF
38.80 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg