Learn More
MP Biomedicals™ Albumin, Bovine Fraction V, fatty acid free, low endotoxin, cell culture reagent
Albumin, Bovine Fraction V, fatty acid free, low endotoxin, cell culture reagent, Off White Shiny Flakes
Brand: MP Biomedicals™ 0219477201
Additional Details : CAS Number : 9048-46-8 Weight : 0.01000kg
Description
- Fraction V
- Cell Culture Reagent
- Fatty Acid Free
- Low endotoxin
- Albumins are a group of acidic proteins which occur in the body fluids and tissues of mammals and in some plant seeds.
- Serum and plasma albumin is carbohydrate free and comprises 55-62% of the protein present. However, only about 40% of the total albumin in the body is in the circulating plasma at one time with the remainder being in extracellular spaces with which there is, in general, equilibration about every 24 hours.
Specifications
∽66338 | |
1g | |
Proteins, Enzymes & Peptides |
6.8-7.2 (7% aq soln) | |
+4°C. | |
Flakes |