Learn More
Miele™ AK12 Half Basket Insert
Brand: Miele™ 3830510
Additional Details : Weight : 1.00000kg
Specifications
442mm | |
Funnels, beakers, wide-mouth glassware, breast pumps, various items | |
AK12 Half Basket Insert |
1Each | |
67mm | |
225mm |