Learn More
Thermo Scientific™ Air Filter Replacement Kits for Ultra Low Temperature Freezers
Easily replace the air filters on your ultra-low temperature freezer with these replacement parts.
Brand:  Thermo Scientific™ 195948
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11800563
  • 139.00 EUR / Pack of 5
Estimated Shipment: 31-05-2024
Description
Easily replace the air filters on your ultra-low temperature freezer with Thermo Scientific™ Air Filter Replacement Kits for Ultra Low Temperature Freezers.
Replacement air filter
Specifications
Specifications
For use with Forma 8600, HERAfreeze HFC, Revco CxF and TSC-Series 12.7, 17.0 and 20.0 cu. ft. Chest ULT Freezers
20 cu. ft.
Chest
Freezer Air Filter
566.3 L
Revco™ CxF, HERAfreeze™ HFC, TSC-Series 12.7, 17.0 and 20.0 cu. ft. Chest ULT Freezers