Learn More
Terumo™ Agani Hypodermic Needles
Used in conjunction with Terumo™ Polypropylene Syringes. Terumo™ Agani Hypodermic Needles are triple-beveled and silicone lubricated for easy insertion.
5.25 EUR - 5.40 EUR
Specifications
Material Stainless steel
Needle Point Style Triple-beveled, 11°
View More Specs
Products 7
Product Code Brand Needle Gauge Diameter (Metric) Outer Length (Metric) Needle Price Quantity & Availability  
Product Code Brand Needle Gauge Diameter (Metric) Outer Length (Metric) Needle Price Quantity & Availability  
15301557 Terumo™
8AN2613R1
26 G 0.45mm 13mm 5.40 EUR
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
08-04-2019
Add to basket
15321547 Terumo™
8AN2238R1
22 G 0.7mm 38mm 5.25 EUR
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
08-04-2019
Add to basket
15341547 Terumo™
8AN2325R1
23 G 0.6mm 25mm 5.25 EUR
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
08-04-2019
Add to basket
15341557 Terumo™
8AN2719R1
27 G 0.4mm 19mm 5.40 EUR
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
08-04-2019
Add to basket
15361537 Terumo™
8AN2038R1
20 G 0.9mm 38mm 5.40 EUR
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
08-04-2019
Add to basket
15371547 Terumo™
8AN2516R1
25 G 0.5mm 16mm 5.25 EUR
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
08-04-2019
Add to basket
15391537 Terumo™
8AN2138R1
21 G 0.8mm 38 mm 5.25 EUR
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
08-04-2019
Add to basket
Description
Description
  • Stainless steel
  • Triple-beveled points
  • Silicone lubrication
Specifications
Specifications
Stainless steel
Triple-beveled, 11°