Learn More
Memmert™ Additional Stainless Steel Grid for IF110plus, Reinforced Chamber Option
Additional grid features a sturdy stainless steel construction.
Brand: Memmert™ IF110plusE29767
Additional Details : Weight : 0.01200kg
Description
- Reinforced
- Stainless Steel
- Electropoloished
- Max. loading 60 kg
Specifications
60 kg | |
IF110plus Incubator | |
Stainless Steel | |
Reinforced Grid |