Learn More
HUBER Adapter for Bath
Accessory for thermostats.
Brand:  HUBER 6391
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 10600693
  • 119.78 EUR / Each
Estimated Shipment: 20-06-2024
Description
  • Adapter
Specifications
Specifications
Thread Size: M30 x 1.5 female to G1/2 male
Adapter