Learn More
HUBER Adapter for Bath
Accessory for thermostats.
Brand:  HUBER 6229
Additional Details : Weight : 0.07200kg
 View more versions of this product
Product Code. 10209511
  • 83.05 EUR / Each
Estimated Shipment: 20-06-2024
Description
  • Adapter
Specifications
Specifications
Thread Size: M16 x 1 female to G 1/2 female
Adapter