Learn More
Eppendorf™ Adapter for 25/50 mL Conical Tubes and Plates
Conical tube and plate adapter for rotor S-4xUniversal-Large.
Brand: Eppendorf™ 5920756001
Additional Details : Weight : 0.72000kg
Description
- 2 pieces
- Adapter
Specifications
Adapter | |
25 mL/50 mL conical tubes and plates | |
For Rotor S-4xUniversal-Large | |
2 |