Learn More
Honeywell™ Adaptec Safety Glasses, Regular
Catalog Number p-7159667
Click to view available options
Lens Tint:
Amber
Clear
Smoke
Designed to fit the widest range of workers, these safety glasses are stylish, comfortable, and adjustable, and feature excellent protection, optics, and Hydroshield anti-fog/anti-scratch coating.
  • Three-position nose bridge is simple to adjust and keeps eyes centered in the lens, where optical clarity is best
  • Workers' vision is less strained so they experience less eye fatigue, work more comfortably for longer periods and can more clearly see their work
  • Soft, flexible material allows each nose pad to independently conform to the wearer's face, delivering greater comfort, contact and stability
  • Adjustable lens angle improves safety and reduces fogging.
  • Each temple hinge ratchets to seven positions - 30 degrees of custom adjustability so lens is angled for both optimal eye protection and ideal space for air circulation
  • A proprietary temple design (patent pending) allows workers to adjust temple tension anytime, safely and without removing eyewear
  • Temple arm features a rigid insert that slides from front to back to control pressure against the head for a firmer or looser fit.
  • Available in regular, narrow, and wide designs
  • One style means simplicity of stocking and ordering as well as consistent eyewear usage by all workers in your facility. No longer is there a need to stock 3-5 different styles of eyewear from multiple manufacturers
  • Vented, cushioned frame for less fogging
  • The dual-action HydroShield anti-fog lens coating provides more than 20X longer lasting fog-free performance - even after extended wear and repeated cleanings and provides 2X greater scratch-resistance
TRUSTED_SUSTAINABILITY
Lens Type Hydroshield coating
Lens Coating Anti-fog, Scratch-resistant
Description Dielectric
Certifications/Compliance (Europe) EC Category PPE: 2, QA: ISO 9001/2000, EC Certificate: C2317.1N, EC Attestation: C2317.1N
Lens Tint Clear
For Use With (Application) Building, construction, general industry, maintenance
Frame Color Red, Black
Lens Material Polycarbonate
Frame Material Polycarbonate