missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACVR2A Human Protein

Easily screen for kinase binding. Thermo Fisher Invitrogen™ ACVR2A Human Protein recombinant fusion protein consists of full-length activin A receptor kinase and a glutathione S-transferase (GST) tag. It is validated in the LanthaScreen™ Eu Kinase Binding Assay.

Brand:  Invitrogen™ PR9111A

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15458713

  • 1826.00 EUR / 100µg
Estimated Shipment: 08-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The LanthaScreen™ Eu Kinase Binding Assay is a TR-FRET assay that lets you measure inhibitor binding to non-activated kinases, perform simple off-rate measurements and makes simple the study of type III (allosteric) inhibitors.

The LanthatScreen™ Eu Kinase Binding Assay is easy to optimize and run. The assay uses a simple mix-and-read format with a single one-hour incubation and no development steps. Simultaneous binding of an Alexa Fluor™ conjugate or tracer” to a tagged kinase is detected by addition of a europium-labeled anti-tag antibody, in this case anti-GST. Simultaneous binding of the tracer and antibody to the kinase results in a high degree of fluorescence resonance energy transfer (FRET), while displacement of the tracer with a kinase inhibitor results in a loss of FRET.

  • Compatible and validated with the LanthaScreen™ Eu Kinase Binding Assay
  • Expressed in insect cells
  • GST tagged for use with Eu-labeled GST antibody
  • No special measures were taken to activate this kinase

ACVR2A for the LanthaScreen Kinase Binding Assay
Activin A receptor kinase (ACVR2A) is a type II member of the TGF-beta family of receptor Serine/Threonine kinases. ACVR2A is involved in the activin and myostatin signaling pathway. (ACVR2A Aliases: ACVR2, ACTRII).

This insect cell-expressed recombinant fusion protein of full-length ACVR2A and GST tag has been validated in the LanthaScreen™ Eu Kinase Binding Assay with the Tracer 236 and Eu-labeled GST reagents.

Specifications

Specifications

NP001607.1
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% NP-40, 2mM DTT and 50% glycerol.
100μg
RUO - research use only
ACVR2A, ACTR2, ACTRII, ACVR2
Human
GST Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
85.3kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
STKR Kinase Family
Recombinant
Baculovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers