Learn More
Cytiva ActiSM liquid medium
A chemically defined ADCF medium optimized for high-yield protein production. Designed for use with ActiPro medium.
Brand: Cytiva SH31040.01
Additional Details : Weight : 0.50000kg
Description
- HyClone ActiSM cell culture medium has been formulated to provide high yields of recombinant proteins in cell culture processes when using Chinese hamster ovary (CHO) cell lines.
- The medium is chemically defined, animal-derived component-free (ADCF), and optimized for high-yield protein production.
- ActiSM medium does not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol, ensuring batch-to-batc
- ActiSM medium is intended for use in combination with HyClone ActiPro medium and Cell Boost™ 7a and Cell Boost 7b supplements as part of the ActiPro system of media and supplements designed with adaptation and production in mind.
- The ActiPro system is suitable for general biomanufacturing with CHO cell lines such as CHO-GS, CHO-K1, CHO-DG44, and CHO-S.
Spécification
2 to 8 °C, away from light | |
Sterile | |
Poloxamer 188 | |
Bottle |
Liquid | |
Sterile-filtered | |
No Glutamine | |
500 mL |