Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Aqueous Filtration
Optimized for a variety of applications requiring larger per well volume processing.
Brand: Cytiva (formerly Pall Lab) 8027
Additional Details : Weight : 1.90000kg
Description
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Polypropylene/Polyethylene Non-woven Media | |
Non-sterile | |
10/Pk. |
30 to 40 μm | |
Polypropylene Housing | |
350 μL |